Assay Details
Gene Symbol: C17orf77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001302809.1 | NP_001289738.1 | 3 | 842 | 108 | |
NM_152460.2 | NP_689673.2 | 3 | 678 | 108 | ||
GenBank mRNA | AK056444.1 | - | 3 | 678 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1700011703.hg.1 | ENST00000375352.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011703.hg.1 | ENST00000375352.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011703.hg.1 | ENST00000375352.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011703.hg.1 | ENST00000375352.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011703.hg.1 | ENST00000375352.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011703.hg.1 | ENST00000375352.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011703.hg.1 | ENST00000375352.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011703.hg.1 | ENST00000375352.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011703.hg.1 | NM_001115152.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011703.hg.1 | NM_001115152.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011703.hg.1 | NM_001115152.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011703.hg.1 | NM_001115152.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011703.hg.1 | NM_001115152.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011703.hg.1 | NM_001115152.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | ENST00000328023.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | ENST00000328023.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | ENST00000328023.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | ENST00000328023.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | ENST00000328023.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | ENST00000392620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | ENST00000392620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | ENST00000392620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | ENST00000392620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | ENST00000392620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | ENST00000392620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | ENST00000524389.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | ENST00000524389.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | ENST00000524389.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | ENST00000524389.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | NM_001302809.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | NM_001302809.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | NM_001302809.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | NM_001302809.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | NM_001302809.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | NM_001302809.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | NM_152460.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | NM_152460.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | NM_152460.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | NM_152460.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | NM_152460.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700008859.hg.1 | NM_152460.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | ENST00000328023.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | ENST00000328023.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | ENST00000328023.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | ENST00000328023.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | ENST00000328023.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | ENST00000392620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | ENST00000392620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | ENST00000392620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | ENST00000392620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | ENST00000392620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | ENST00000392620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | ENST00000524389.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | ENST00000524389.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | ENST00000524389.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | ENST00000524389.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | NM_001302809.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | NM_001302809.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | NM_001302809.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | NM_001302809.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | NM_001302809.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | NM_001302809.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | NM_152460.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | NM_152460.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | NM_152460.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | NM_152460.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | NM_152460.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700008859.hg.1 | NM_152460.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011703.hg.1 | ENST00000375352.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011703.hg.1 | ENST00000375352.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011703.hg.1 | ENST00000375352.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011703.hg.1 | ENST00000375352.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011703.hg.1 | ENST00000375352.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011703.hg.1 | ENST00000375352.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011703.hg.1 | ENST00000375352.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011703.hg.1 | ENST00000375352.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011703.hg.1 | NM_001115152.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011703.hg.1 | NM_001115152.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011703.hg.1 | NM_001115152.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011703.hg.1 | NM_001115152.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011703.hg.1 | NM_001115152.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011703.hg.1 | NM_001115152.2 | Refseq |
