Assay Details
| Gene Symbol: RNF152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173557.2 | NP_775828.1 | 2 | 1015 | 70 | |
| XM_005266650.3 | XP_005266707.1 | 4 | 1216 | 70 | ||
| XM_005266652.3 | XP_005266709.1 | 2 | 1009 | 70 | ||
| XM_011525878.2 | XP_011524180.1 | 4 | 1347 | 70 | ||
| XM_011525879.2 | XP_011524181.1 | 5 | 1527 | 70 | ||
| XM_017025612.1 | XP_016881101.1 | 3 | 1132 | 70 | ||
| XM_017025613.1 | XP_016881102.1 | 4 | 1333 | 70 | ||
| GenBank mRNA | AK096495.1 | - | 2 | 1015 | 70 | |
| AK122758.1 | - | 2 | 1703 | 70 | ||
| BC094004.1 | - | 1 | 625 | 7939849 | 70 | |
| BC111956.1 | - | 1 | 625 | 8327424 | 70 | |
More Information
Set Membership: |
|

RNF152
Genomic Map