| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000269489.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000269489.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000269489.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000269489.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000269489.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000269489.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000269489.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000269489.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000269489.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000269489.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000269489.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000269489.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000269489.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000269489.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000269489.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000344731.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000344731.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000344731.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000344731.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000344731.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000344731.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000344731.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000344731.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000344731.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000344731.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000344731.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000344731.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000344731.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000415733.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000415733.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000415733.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000415733.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000431153.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000431153.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000431153.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000431153.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000431153.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000431153.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000438844.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000438844.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000438844.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000438844.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000438844.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000438844.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000438844.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000438844.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000438844.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000438844.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000438844.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000438844.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000479842.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000479842.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000479842.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000479842.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
ENST00000479842.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
NM_001307923.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
NM_001307923.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
NM_001307923.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
NM_001307923.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
NM_001307923.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
NM_001307923.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
NM_001307923.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
NM_001307923.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
NM_001307923.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
NM_001307923.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
NM_001307923.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
NM_001307923.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
NM_001307923.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
NM_001307923.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
NM_001307923.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
NM_012397.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
NM_012397.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
NM_012397.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
NM_012397.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
NM_012397.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
NM_012397.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
NM_012397.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
NM_012397.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
NM_012397.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
NM_012397.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
NM_012397.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
NM_012397.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
NM_012397.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800007547.hg.1 |
NM_012397.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000269489.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000269489.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000269489.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000269489.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000269489.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000269489.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000269489.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000269489.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000269489.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000269489.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000269489.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000269489.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000269489.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000269489.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000269489.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000344731.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000344731.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000344731.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000344731.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000344731.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000344731.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000344731.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000344731.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000344731.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000344731.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000344731.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000344731.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000344731.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000415733.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000415733.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000415733.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000415733.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000431153.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000431153.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000431153.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000431153.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000431153.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000431153.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000438844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000438844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000438844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000438844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000438844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000438844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000438844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000438844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000438844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000438844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000438844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000438844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000479842.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000479842.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000479842.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000479842.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
ENST00000479842.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
NM_001307923.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
NM_001307923.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
NM_001307923.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
NM_001307923.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
NM_001307923.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
NM_001307923.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
NM_001307923.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
NM_001307923.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
NM_001307923.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
NM_001307923.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
NM_001307923.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
NM_001307923.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
NM_001307923.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
NM_001307923.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
NM_001307923.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
NM_012397.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
NM_012397.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
NM_012397.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
NM_012397.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
NM_012397.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
NM_012397.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
NM_012397.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
NM_012397.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
NM_012397.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
NM_012397.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
NM_012397.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
NM_012397.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
NM_012397.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800007547.hg.1 |
NM_012397.3 |
Refseq |