Assay Details
Gene Symbol: RNF165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK091537.1 | - | 1 | 1366 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000587853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000587853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000587853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000587853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000587853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000587853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000587853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000588679.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000588679.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000588679.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000588679.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000588679.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000588679.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000588679.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000590330.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000590330.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000590330.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000590330.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000590330.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000592777.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000592777.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000592777.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000592777.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000592777.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000592777.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000592777.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000593230.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000593230.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000593230.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000593230.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000593230.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | ENST00000593230.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NR_046357.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NR_046357.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NR_046357.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NR_046357.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007220.hg.1 | NR_046357.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000269439.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000543885.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000586604.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000587853.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000587853.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000587853.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000587853.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000587853.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000587853.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000587853.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000588679.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000588679.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000588679.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000588679.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000588679.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000588679.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000588679.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000590330.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000590330.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000590330.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000590330.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000590330.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000592777.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000592777.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000592777.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000592777.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000592777.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000592777.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000592777.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000593230.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000593230.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000593230.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000593230.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000593230.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | ENST00000593230.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_001256758.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NM_152470.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NR_046357.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NR_046357.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NR_046357.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NR_046357.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007220.hg.1 | NR_046357.1 | Refseq |
