Assay Details
| Gene Symbol: LOC100505635 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK091770.1 | - | 1 | 2814 | 82 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000420293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000420293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000420293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000420293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000420293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000420293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000437947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000437947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000437947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000606829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000606829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000606829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000606829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000606829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000607215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000607215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000607215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000607215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000607675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000607675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000607675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000607675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000607675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000607675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000607675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | NR_125831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | NR_125831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | NR_125831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | NR_125831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | NR_125831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | NR_125831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | NR_125831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | NR_125831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000420293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000420293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000420293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000420293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000420293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000420293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000437947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000437947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000437947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000606829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000606829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000606829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000606829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000606829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000607215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000607215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000607215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000607215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000607675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000607675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000607675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000607675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000607675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000607675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000607675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | NR_125831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | NR_125831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | NR_125831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | NR_125831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | NR_125831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | NR_125831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | NR_125831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | NR_125831.1 | Refseq |

LOC100505635
Genomic Map