Assay Details
| Gene Symbol: STK19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004197.1 | NP_004188.1 | 8 | 1498 | 73 | |
| NM_032454.1 | NP_115830.1 | 8 | 1510 | 73 | ||
| NR_026717.1 | 8 | 1683 | 73 | |||
| GenBank mRNA | BC110347.1 | - | 8 | 1683 | 5165123 | 73 |
| L26260.1 | - | 8 | 1498 | 73 | ||
| X77386.1 | - | 7 | 1104 | 73 | ||
More Information
Set Membership: |
|

STK19
Genomic Map