Assay Details
| Gene Symbol: MYB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130172.1 | NP_001123644.1 | 5 | 625 | 96 | |
| NM_001130173.1 | NP_001123645.1 | 5 | 625 | 96 | ||
| NM_001161656.1 | NP_001155128.1 | 5 | 625 | 96 | ||
| NM_001161657.1 | NP_001155129.1 | 5 | 625 | 96 | ||
| NM_001161658.1 | NP_001155130.1 | 5 | 625 | 96 | ||
| NM_001161659.1 | NP_001155131.1 | 5 | 625 | 96 | ||
| NM_001161660.1 | NP_001155132.1 | 5 | 625 | 96 | ||
| NM_005375.3 | NP_005366.2 | 5 | 625 | 96 | ||
| NR_134958.1 | 5 | 625 | 96 | |||
| NR_134959.1 | 5 | 625 | 96 | |||
| NR_134960.1 | 5 | 625 | 96 | |||
| NR_134961.1 | 5 | 625 | 96 | |||
| NR_134963.1 | 5 | 625 | 96 | |||
| NR_134964.1 | 5 | 625 | 96 | |||
| NR_134965.1 | 5 | 625 | 96 | |||
| GenBank mRNA | AF104863.1 | - | 5 | 503 | 96 | |
| AJ606317.1 | - | 5 | 625 | 96 | ||
| AJ606318.1 | - | 5 | 625 | 96 | ||
| AJ606319.1 | - | 5 | 625 | 96 | ||
| AJ606320.1 | - | 5 | 625 | 96 | ||
| AJ606321.1 | - | 5 | 625 | 96 | ||
| AJ606322.1 | - | 5 | 625 | 96 | ||
| AJ606323.1 | - | 5 | 625 | 96 | ||
| AJ606324.1 | - | 5 | 625 | 96 | ||
| AJ616235.1 | - | 5 | 540 | 96 | ||
| AJ616795.1 | - | 1 | 120 | 96 | ||
| AY787443.1 | - | 5 | 426 | 96 | ||
| AY787445.1 | - | 4 | 354 | 96 | ||
| AY787447.1 | - | 5 | 426 | 96 | ||
| AY787448.1 | - | 5 | 426 | 96 | ||
| AY787449.1 | - | 5 | 426 | 96 | ||
| AY787450.1 | - | 5 | 426 | 96 | ||
| AY787451.1 | - | 5 | 426 | 96 | ||
| AY787452.1 | - | 5 | 426 | 96 | ||
| AY787453.1 | - | 5 | 426 | 96 | ||
| AY787454.1 | - | 5 | 426 | 96 | ||
| AY787455.1 | - | 5 | 426 | 96 | ||
| AY787456.1 | - | 5 | 426 | 96 | ||
| AY787457.1 | - | 5 | 426 | 96 | ||
| AY787458.1 | - | 5 | 426 | 96 | ||
| AY787459.1 | - | 5 | 426 | 96 | ||
| AY787460.1 | - | 5 | 426 | 96 | ||
| AY787461.1 | - | 5 | 426 | 96 | ||
| AY787462.1 | - | 5 | 426 | 96 | ||
| AY787463.1 | - | 5 | 426 | 96 | ||
| AY787464.1 | - | 5 | 426 | 96 | ||
| AY787465.1 | - | 5 | 426 | 96 | ||
| AY787466.1 | - | 5 | 426 | 96 | ||
| AY787467.1 | - | 5 | 426 | 96 | ||
| AY787468.1 | - | 5 | 426 | 96 | ||
| AY787469.1 | - | 5 | 426 | 96 | ||
| AY787470.1 | - | 5 | 426 | 96 | ||
| AY787471.1 | - | 5 | 426 | 96 | ||
| AY787472.1 | - | 5 | 426 | 96 | ||
| AY787473.1 | - | 5 | 426 | 96 | ||
| AY787474.1 | - | 5 | 426 | 96 | ||
| AY787475.1 | - | 5 | 426 | 96 | ||
| BC064955.1 | - | 5 | 577 | 6069320 | 96 | |
| M13665.1 | - | 3 | 288 | 96 | ||
| M13666.1 | - | 3 | 288 | 96 | ||
| M15024.1 | - | 5 | 539 | 96 | ||
| X52125.1 | - | 5 | 623 | 96 | ||
| X52126.1 | - | 4 | 429 | 96 | ||
More Information
Set Membership: |
|

MYB
Genomic Map