Assay Details
| Gene Symbol: SNX9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005267015.2 | XP_005267072.1 | 2 - 3 | 311 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005267015.2 transcripts.
Set Membership: |
|

SNX9
Genomic Map