Assay Details
| Gene Symbol: BLOC1S5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK311385.1 | - | 1 - 2 | 266 | 93 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | ENST00000244777.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | ENST00000244777.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | ENST00000244777.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | ENST00000244777.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | ENST00000244777.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | ENST00000244777.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | ENST00000244777.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | ENST00000244777.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | ENST00000397457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | ENST00000397457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | ENST00000397457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | ENST00000397457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | ENST00000397457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | ENST00000397457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | ENST00000397457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | ENST00000397457.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | ENST00000475998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | ENST00000475998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | ENST00000475998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | ENST00000486432.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | ENST00000486432.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | ENST00000486432.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | ENST00000486432.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | ENST00000543936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | ENST00000543936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | ENST00000543936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | ENST00000543936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | ENST00000543936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | ENST00000543936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | ENST00000543936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | ENST00000627748.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | ENST00000627748.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | ENST00000627748.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | ENST00000627748.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | ENST00000627748.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | ENST00000627748.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | ENST00000627748.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | NM_001199322.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | NM_001199322.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | NM_001199322.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | NM_001199322.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | NM_001199322.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | NM_001199322.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | NM_001199322.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | NM_001199322.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | NM_001199322.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | NM_001199323.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | NM_001199323.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | NM_001199323.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | NM_001199323.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | NM_001199323.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | NM_001199323.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | NM_001199323.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | NM_201280.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | NM_201280.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | NM_201280.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | NM_201280.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | NM_201280.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | NM_201280.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | NM_201280.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014238.hg.1 | NM_201280.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | ENST00000244777.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | ENST00000244777.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | ENST00000244777.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | ENST00000244777.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | ENST00000244777.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | ENST00000244777.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | ENST00000244777.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | ENST00000244777.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | ENST00000397457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | ENST00000397457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | ENST00000397457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | ENST00000397457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | ENST00000397457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | ENST00000397457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | ENST00000397457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | ENST00000397457.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | ENST00000475998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | ENST00000475998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | ENST00000475998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | ENST00000486432.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | ENST00000486432.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | ENST00000486432.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | ENST00000486432.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | ENST00000543936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | ENST00000543936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | ENST00000543936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | ENST00000543936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | ENST00000543936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | ENST00000543936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | ENST00000543936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | ENST00000627748.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | ENST00000627748.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | ENST00000627748.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | ENST00000627748.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | ENST00000627748.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | ENST00000627748.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | ENST00000627748.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | NM_001199322.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | NM_001199322.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | NM_001199322.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | NM_001199322.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | NM_001199322.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | NM_001199322.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | NM_001199322.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | NM_001199322.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | NM_001199322.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | NM_001199323.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | NM_001199323.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | NM_001199323.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | NM_001199323.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | NM_001199323.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | NM_001199323.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | NM_001199323.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | NM_201280.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | NM_201280.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | NM_201280.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | NM_201280.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | NM_201280.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | NM_201280.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | NM_201280.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014238.hg.1 | NM_201280.2 | Refseq |

BLOC1S5
Genomic Map