Assay Details
| Gene Symbol: ZNF273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017011693.1 | XP_016867182.1 | 2 - 3 | 339 | 99 | |
| GenBank mRNA | AK090648.1 | - | 2 - 3 | 339 | 99 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK090648.1, XM_017011693.1 transcripts.
Set Membership: |
|

ZNF273
Genomic Map