Assay Details
| Gene Symbol: ZNF273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017011689.1 | XP_016867178.1 | 4 - 5 | 5394 | 96 | |
| XM_017011690.1 | XP_016867179.1 | 3 - 4 | 5305 | 96 | ||
| XM_017011691.1 | XP_016867180.1 | 4 - 5 | 2596 | 96 | ||
| XM_017011693.1 | XP_016867182.1 | 3 - 4 | 486 | 96 | ||
| XM_017011697.1 | XP_016867186.1 | 3 - 4 | 5305 | 96 | ||
| XM_017011698.1 | XP_016867187.1 | 2 - 3 | 422 | 96 | ||
| GenBank mRNA | AK090648.1 | - | 3 - 4 | 486 | 96 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK090648.1, XM_017011689.1, XM_017011690.1, XM_017011691.1, XM_017011693.1, XM_017011697.1, XM_017011698.1 transcripts.
Set Membership: |
|

ZNF273
Genomic Map