| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012301.hg.1 |
ENST00000397764.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012301.hg.1 |
ENST00000397764.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012301.hg.1 |
ENST00000397764.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012301.hg.1 |
ENST00000397764.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012301.hg.1 |
ENST00000397764.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012301.hg.1 |
ENST00000397764.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012301.hg.1 |
ENST00000397764.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012301.hg.1 |
ENST00000413744.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012301.hg.1 |
ENST00000413744.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012301.hg.1 |
ENST00000413744.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012301.hg.1 |
ENST00000413744.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012301.hg.1 |
ENST00000413744.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012301.hg.1 |
ENST00000439551.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012301.hg.1 |
ENST00000439551.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012301.hg.1 |
ENST00000439551.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012301.hg.1 |
ENST00000439551.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012301.hg.1 |
ENST00000441359.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012301.hg.1 |
ENST00000441359.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012301.hg.1 |
ENST00000441359.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012301.hg.1 |
ENST00000441359.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012301.hg.1 |
ENST00000441359.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012301.hg.1 |
ENST00000441359.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012301.hg.1 |
NR_015442.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012301.hg.1 |
NR_015442.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012301.hg.1 |
NR_015442.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012301.hg.1 |
NR_015442.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012301.hg.1 |
NR_015442.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012301.hg.1 |
NR_015442.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012301.hg.1 |
NR_024412.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012301.hg.1 |
NR_024412.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012301.hg.1 |
NR_024412.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012301.hg.1 |
NR_024412.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012301.hg.1 |
NR_024412.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012301.hg.1 |
NR_024412.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012301.hg.1 |
NR_024412.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012301.hg.1 |
ENST00000397764.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012301.hg.1 |
ENST00000397764.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012301.hg.1 |
ENST00000397764.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012301.hg.1 |
ENST00000397764.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012301.hg.1 |
ENST00000397764.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012301.hg.1 |
ENST00000397764.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012301.hg.1 |
ENST00000397764.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012301.hg.1 |
ENST00000413744.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012301.hg.1 |
ENST00000413744.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012301.hg.1 |
ENST00000413744.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012301.hg.1 |
ENST00000413744.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012301.hg.1 |
ENST00000413744.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012301.hg.1 |
ENST00000439551.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012301.hg.1 |
ENST00000439551.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012301.hg.1 |
ENST00000439551.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012301.hg.1 |
ENST00000439551.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012301.hg.1 |
ENST00000441359.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012301.hg.1 |
ENST00000441359.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012301.hg.1 |
ENST00000441359.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012301.hg.1 |
ENST00000441359.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012301.hg.1 |
ENST00000441359.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012301.hg.1 |
ENST00000441359.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012301.hg.1 |
NR_015442.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012301.hg.1 |
NR_015442.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012301.hg.1 |
NR_015442.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012301.hg.1 |
NR_015442.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012301.hg.1 |
NR_015442.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012301.hg.1 |
NR_015442.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012301.hg.1 |
NR_024412.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012301.hg.1 |
NR_024412.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012301.hg.1 |
NR_024412.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012301.hg.1 |
NR_024412.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012301.hg.1 |
NR_024412.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012301.hg.1 |
NR_024412.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012301.hg.1 |
NR_024412.1 |
Refseq |