Assay Details
| Gene Symbol: LINC01510 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120506.1 | 1 - 2 | 222 | 65 | ||
| GenBank mRNA | DR422469.1 | - | 1 - 2 | 222 | 65 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000421965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000421965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000421965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000421965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000421965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000421965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000441991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000441991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000441991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000441991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000441991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000450063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000450063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000450063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000450063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000458082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000458082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000458082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000458082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000458082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000458082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | ENST00000458082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | NR_120506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | NR_120506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | NR_120506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | NR_120506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | NR_120506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | NR_120506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | NR_120506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | NR_120506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | NR_120506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | NR_120506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | NR_120506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | NR_120506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | NR_120506.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012344.hg.1 | NR_120506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000421965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000421965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000421965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000421965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000421965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000421965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000441991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000441991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000441991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000441991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000441991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000450063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000450063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000450063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000450063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000458082.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000458082.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000458082.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000458082.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000458082.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000458082.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | ENST00000458082.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | NR_120506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | NR_120506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | NR_120506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | NR_120506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | NR_120506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | NR_120506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | NR_120506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | NR_120506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | NR_120506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | NR_120506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | NR_120506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | NR_120506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | NR_120506.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012344.hg.1 | NR_120506.1 | Refseq |

LINC01510
Genomic Map