Assay Details
| Gene Symbol: ZNF800 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006715867.3 | XP_006715930.1 | 5 - 6 | 2790 | 93 | |
| XM_006715868.3 | XP_006715931.1 | 5 - 6 | 2785 | 93 | ||
| XM_006715869.3 | XP_006715932.1 | 4 - 5 | 2694 | 93 | ||
| XM_011515859.2 | XP_011514161.1 | 5 - 6 | 2506 | 93 | ||
More Information
Set Membership: |
|

ZNF800
Genomic Map