Assay Details
| Gene Symbol: PARP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022750.3 | NP_073587.1 | 10 - 11 | 2500 | 63 | |
| NR_130117.1 | 9 - 10 | 2358 | 63 | |||
| GenBank mRNA | AK026346.1 | - | 4 - 5 | 340 | 63 | |
| AL136766.1 | - | 10 - 11 | 2122 | 63 | ||
| AL137255.1 | - | 10 - 11 | 1319 | 63 | ||
| BC081541.1 | - | 10 - 11 | 2500 | 5296560 | 63 | |
More Information
Set Membership: |
|

PARP12
Genomic Map