Assay Details
| Gene Symbol: PMS2P4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | D38438.1 | - | 6 - 7 | 610 | 80 | |
| D38500.1 | - | 6 - 7 | 610 | 80 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PMS2P4| Gene Symbol: PMS2P4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | D38438.1 | - | 6 - 7 | 610 | 80 | |
| D38500.1 | - | 6 - 7 | 610 | 80 | ||
Set Membership: |
|