| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009187.hg.1 |
ENST00000525260.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009187.hg.1 |
ENST00000525260.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009187.hg.1 |
ENST00000525260.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009187.hg.1 |
ENST00000525260.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009187.hg.1 |
ENST00000525260.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009187.hg.1 |
ENST00000525260.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009187.hg.1 |
ENST00000534150.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009187.hg.1 |
ENST00000534150.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009187.hg.1 |
ENST00000534150.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009187.hg.1 |
ENST00000534150.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009187.hg.1 |
ENST00000534150.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009187.hg.1 |
ENST00000534150.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009187.hg.1 |
ENST00000534150.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009187.hg.1 |
ENST00000534150.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009187.hg.1 |
ENST00000581173.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009187.hg.1 |
ENST00000581173.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009187.hg.1 |
ENST00000581173.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009187.hg.1 |
ENST00000581173.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009187.hg.1 |
ENST00000525260.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009187.hg.1 |
ENST00000525260.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009187.hg.1 |
ENST00000525260.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009187.hg.1 |
ENST00000525260.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009187.hg.1 |
ENST00000525260.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009187.hg.1 |
ENST00000525260.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009187.hg.1 |
ENST00000534150.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009187.hg.1 |
ENST00000534150.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009187.hg.1 |
ENST00000534150.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009187.hg.1 |
ENST00000534150.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009187.hg.1 |
ENST00000534150.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009187.hg.1 |
ENST00000534150.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009187.hg.1 |
ENST00000534150.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009187.hg.1 |
ENST00000534150.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009187.hg.1 |
ENST00000581173.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009187.hg.1 |
ENST00000581173.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009187.hg.1 |
ENST00000581173.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009187.hg.1 |
ENST00000581173.2 |
Ensembl |