Assay Details
| Gene Symbol: LOC100128239 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC043590.1 | - | 1 - 2 | 275 | 92 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000529070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000529070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000529070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000529070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000529070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000529070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000529070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000529070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000531938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000531938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000531938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000531938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000531938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000532327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000532327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000532327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000532327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000532327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000532327.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000532706.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000532706.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000532706.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000532706.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000529070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000529070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000529070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000529070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000529070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000529070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000529070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000529070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000531938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000531938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000531938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000531938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000531938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000532327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000532327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000532327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000532327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000532327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000532327.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000532706.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000532706.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000532706.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000532706.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | ENST00000533922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | NR_027276.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013116.hg.1 | NR_027276.1 | Refseq |

LOC100128239
Genomic Map