Assay Details
| Gene Symbol: ACTR3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017012548.1 | XP_016868037.1 | 15 | 4559 | 97 | |
| XM_017012550.1 | XP_016868039.1 | 11 | 7254 | 97 | ||
| XM_017012551.1 | XP_016868040.1 | 10 | 7129 | 97 | ||
More Information
Set Membership: |
|

ACTR3C
Genomic Map