| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010568.hg.1 |
ENST00000222753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010568.hg.1 |
ENST00000222753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010568.hg.1 |
ENST00000222753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010568.hg.1 |
ENST00000222753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010568.hg.1 |
ENST00000222753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010568.hg.1 |
ENST00000222753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010568.hg.1 |
ENST00000222753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010568.hg.1 |
ENST00000222753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010568.hg.1 |
ENST00000222753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010568.hg.1 |
ENST00000222753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010568.hg.1 |
ENST00000222753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010568.hg.1 |
ENST00000518136.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010568.hg.1 |
ENST00000518136.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010568.hg.1 |
NM_000522.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010568.hg.1 |
NM_000522.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010568.hg.1 |
NM_000522.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010568.hg.1 |
NM_000522.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010568.hg.1 |
NM_000522.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010568.hg.1 |
NM_000522.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010568.hg.1 |
ENST00000222753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010568.hg.1 |
ENST00000222753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010568.hg.1 |
ENST00000222753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010568.hg.1 |
ENST00000222753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010568.hg.1 |
ENST00000222753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010568.hg.1 |
ENST00000222753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010568.hg.1 |
ENST00000222753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010568.hg.1 |
ENST00000222753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010568.hg.1 |
ENST00000222753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010568.hg.1 |
ENST00000222753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010568.hg.1 |
ENST00000222753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010568.hg.1 |
ENST00000518136.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010568.hg.1 |
ENST00000518136.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010568.hg.1 |
NM_000522.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010568.hg.1 |
NM_000522.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010568.hg.1 |
NM_000522.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010568.hg.1 |
NM_000522.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010568.hg.1 |
NM_000522.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010568.hg.1 |
NM_000522.4 |
Refseq |