Assay Details
| Gene Symbol: ABCB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF345626.1 | - | 125 | 78 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ABCB1| Gene Symbol: ABCB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF345626.1 | - | 125 | 78 | ||