Assay Details
| Gene Symbol: PCMTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286782.1 | NP_001273711.1 | 4 | 3370 | 97 | |
| NM_001286783.1 | NP_001273712.1 | 6 | 3665 | 97 | ||
| NM_052937.3 | NP_443169.2 | 6 | 3875 | 97 | ||
| XM_011517444.2 | XP_011515746.1 | 5 | 3464 | 97 | ||
| XM_017013017.1 | XP_016868506.1 | 6 | 3708 | 97 | ||
| GenBank mRNA | AF293366.1 | - | 1 | 97 | ||
| AL833027.1 | - | 5 | 3270 | 97 | ||
| BC010693.1 | - | 4 | 3151 | 97 | ||
| BC017677.2 | - | 1 | 1663 | 97 | ||
| BC032670.1 | - | 5 | 3613 | 5539318 | 97 | |
| BX640885.1 | - | 4 | 3179 | 97 | ||
More Information
Set Membership: |
|

PCMTD1
Genomic Map