Assay Details
| Gene Symbol: RAB11FIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002814.2 | NP_001002814.2 | 6 | 7631 | 87 | |
| NM_025151.4 | NP_079427.4 | 5 | 5729 | 87 | ||
| GenBank mRNA | AK122583.1 | - | 3 | 5772 | 87 | |
| AK226111.1 | - | 6 | 7321 | 87 | ||
More Information
Set Membership: |
|

RAB11FIP1
Genomic Map