Assay Details
| Gene Symbol: LINC01151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_126348.1 | 3 | 387 | 73 | ||
| GenBank mRNA | DB186869.1 | - | 3 | 387 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01151| Gene Symbol: LINC01151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_126348.1 | 3 | 387 | 73 | ||
| GenBank mRNA | DB186869.1 | - | 3 | 387 | 73 | |
Set Membership: |
|