Assay Details
Gene Symbol: HNF4G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011517516.2 | XP_011515818.1 | 1 - 2 | 398 | 64 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011517516.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
intracellular receptor signaling pathway
steroid hormone mediated signaling pathway
transcription factor activity, sequence-specific DNA binding
steroid hormone receptor activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
zinc ion binding
sequence-specific DNA binding
regulation of transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
intracellular receptor signaling pathway
steroid hormone mediated signaling pathway
transcription factor activity, sequence-specific DNA binding
steroid hormone receptor activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
zinc ion binding
sequence-specific DNA binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396419.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396419.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396419.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396419.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000494318.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000494318.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000494318.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000494318.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | ENST00000494318.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | NM_004133.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | NM_004133.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | NM_004133.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | NM_004133.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | NM_004133.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | NM_004133.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | NM_004133.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | NM_004133.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | NM_004133.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | NM_004133.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | NM_004133.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | NM_004133.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | NM_004133.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | NM_004133.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008002.hg.1 | NM_004133.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000354370.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396419.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396419.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396419.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396419.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000396423.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000494318.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000494318.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000494318.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000494318.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | ENST00000494318.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | NM_004133.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | NM_004133.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | NM_004133.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | NM_004133.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | NM_004133.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | NM_004133.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | NM_004133.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | NM_004133.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | NM_004133.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | NM_004133.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | NM_004133.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | NM_004133.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | NM_004133.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | NM_004133.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008002.hg.1 | NM_004133.4 | Refseq |
