Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011500.hg.1 |
ENST00000519506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011500.hg.1 |
ENST00000519506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011500.hg.1 |
ENST00000519506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011500.hg.1 |
ENST00000519506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011500.hg.1 |
ENST00000519506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011500.hg.1 |
ENST00000522778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011500.hg.1 |
ENST00000522778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011500.hg.1 |
ENST00000522778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011500.hg.1 |
NR_125418.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011500.hg.1 |
NR_125418.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011500.hg.1 |
NR_125418.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011500.hg.1 |
NR_125418.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011500.hg.1 |
NR_125418.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011500.hg.1 |
NR_125418.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011500.hg.1 |
ENST00000519506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011500.hg.1 |
ENST00000519506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011500.hg.1 |
ENST00000519506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011500.hg.1 |
ENST00000519506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011500.hg.1 |
ENST00000519506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011500.hg.1 |
ENST00000522778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011500.hg.1 |
ENST00000522778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011500.hg.1 |
ENST00000522778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011500.hg.1 |
NR_125418.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011500.hg.1 |
NR_125418.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011500.hg.1 |
NR_125418.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011500.hg.1 |
NR_125418.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011500.hg.1 |
NR_125418.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011500.hg.1 |
NR_125418.1 |
Refseq |