Assay Details
| Gene Symbol: MCPH1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX374425.2 | - | 1 - 2 | 37 | 61 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800009420.hg.1 | ENST00000515608.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009420.hg.1 | ENST00000515608.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009420.hg.1 | ENST00000515608.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009420.hg.1 | ENST00000515608.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009420.hg.1 | ENST00000515608.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009420.hg.1 | ENST00000515608.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009420.hg.1 | ENST00000515608.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009420.hg.1 | ENST00000515608.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009420.hg.1 | ENST00000515608.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009420.hg.1 | ENST00000515608.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009420.hg.1 | ENST00000515608.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009420.hg.1 | ENST00000515608.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009420.hg.1 | ENST00000522897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009420.hg.1 | ENST00000522897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009420.hg.1 | ENST00000522897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009420.hg.1 | ENST00000522897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009420.hg.1 | ENST00000525186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009420.hg.1 | ENST00000525186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009420.hg.1 | ENST00000525186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009420.hg.1 | ENST00000525186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009420.hg.1 | ENST00000527490.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009420.hg.1 | ENST00000527490.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009420.hg.1 | ENST00000607368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009420.hg.1 | NR_125386.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009420.hg.1 | NR_125386.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009420.hg.1 | NR_125386.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009420.hg.1 | NR_125386.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009420.hg.1 | NR_125386.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009420.hg.1 | NR_125386.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009420.hg.1 | NR_125386.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009420.hg.1 | ENST00000515608.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009420.hg.1 | ENST00000515608.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009420.hg.1 | ENST00000515608.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009420.hg.1 | ENST00000515608.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009420.hg.1 | ENST00000515608.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009420.hg.1 | ENST00000515608.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009420.hg.1 | ENST00000515608.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009420.hg.1 | ENST00000515608.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009420.hg.1 | ENST00000515608.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009420.hg.1 | ENST00000515608.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009420.hg.1 | ENST00000515608.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009420.hg.1 | ENST00000515608.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009420.hg.1 | ENST00000522897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009420.hg.1 | ENST00000522897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009420.hg.1 | ENST00000522897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009420.hg.1 | ENST00000522897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009420.hg.1 | ENST00000525186.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009420.hg.1 | ENST00000525186.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009420.hg.1 | ENST00000525186.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009420.hg.1 | ENST00000525186.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009420.hg.1 | ENST00000527490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009420.hg.1 | ENST00000527490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009420.hg.1 | ENST00000607368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009420.hg.1 | NR_125386.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009420.hg.1 | NR_125386.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009420.hg.1 | NR_125386.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009420.hg.1 | NR_125386.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009420.hg.1 | NR_125386.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009420.hg.1 | NR_125386.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009420.hg.1 | NR_125386.1 | Refseq |

MCPH1-AS1
Genomic Map