Assay Details
| Gene Symbol: LSM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014462.2 | NP_055277.1 | 2 - 3 | 523 | 92 | |
| NR_045493.1 | 2 - 3 | 245 | 92 | |||
| GenBank mRNA | AF000177.1 | - | 2 - 3 | 277 | 92 | |
| AF147348.1 | - | 2 - 3 | 166 | 92 | ||
| AJ238094.1 | - | 2 - 3 | 301 | 92 | ||
| AK312159.1 | - | 2 - 3 | 276 | 92 | ||
| BC001767.1 | - | 2 - 3 | 210 | 3353833 | 92 | |
| BP376421.1 | - | 2 - 3 | 486 | 92 | ||
| BU941962.1 | - | 2 - 3 | 248 | 92 | ||
| KU178606.1 | - | 2 - 3 | 113 | 92 | ||
More Information
Set Membership: |
|

LSM1
Genomic Map