Assay Details
| Gene Symbol: SLC20A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006716391.3 | XP_006716454.1 | 2 - 3 | 4068 | 106 | |
| XM_017013750.1 | XP_016869239.1 | 2 - 3 | 723 | 106 | ||
| XM_017013751.1 | XP_016869240.1 | 2 - 3 | 597 | 106 | ||
| XM_017013752.1 | XP_016869241.1 | 2 - 3 | 839 | 106 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006716391.3, XM_017013750.1, XM_017013751.1, XM_017013752.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transport
ion transport
phosphate ion transmembrane transport
sodium ion transmembrane transport
sodium-dependent phosphate transport
viral entry into host cell
virus receptor activity
receptor activity
inorganic phosphate transmembrane transporter activity
sodium:phosphate symporter activity
sodium:inorganic phosphate symporter activity
sodium-dependent phosphate transmembrane transporter activity
transport
ion transport
phosphate ion transmembrane transport
sodium ion transmembrane transport
sodium-dependent phosphate transport
viral entry into host cell
virus receptor activity
receptor activity
inorganic phosphate transmembrane transporter activity
sodium:phosphate symporter activity
sodium:inorganic phosphate symporter activity
sodium-dependent phosphate transmembrane transporter activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000342228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000342228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000342228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000342228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000342228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000342228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000342228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000342228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000342228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000342228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000342228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000342228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000342228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000342228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000342228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000342228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000342228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000342228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000342228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000517366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000517366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000517366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000517366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000517366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000517366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000518384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000518384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000518384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000518384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000518384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000518660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000518660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000518660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000518660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000518660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000518660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000518717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000518717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000518717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000518717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000518717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000518717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000519463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000519463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000519463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000521455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000521455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000521455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000521455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000522401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000522401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000522401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000522401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000522707.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000522707.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000522707.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000522707.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000522707.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000523340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000523340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000523340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000523340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000523340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000523340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000523340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000523340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000523340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000524211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000524211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000524211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000524211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000524211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000524211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000524211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000524211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000524211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000524237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000524237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000524237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000524237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000524237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |

SLC20A2
Genomic Map