Assay Details
| Gene Symbol: SLC20A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006716391.3 | XP_006716454.1 | 3 - 4 | 4205 | 109 | |
| XM_017013750.1 | XP_016869239.1 | 3 - 4 | 860 | 109 | ||
| XM_017013751.1 | XP_016869240.1 | 3 - 4 | 734 | 109 | ||
| XM_017013752.1 | XP_016869241.1 | 3 - 4 | 976 | 109 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006716391.3, XM_017013750.1, XM_017013751.1, XM_017013752.1 transcripts.
Set Membership: |
|

SLC20A2
Genomic Map