Assay Details
| Gene Symbol: SQLE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003129.3 | NP_003120.2 | 7 - 8 | 2132 | 83 | |
| XM_011517246.2 | XP_011515548.1 | 7 - 8 | 1230 | 83 | ||
| GenBank mRNA | AF098865.1 | - | 7 - 8 | 1361 | 83 | |
| AK225085.1 | - | 7 - 8 | 1422 | 83 | ||
| AK295302.1 | - | 7 - 8 | 1032 | 83 | ||
| AK301792.1 | - | 7 - 8 | 1024 | 83 | ||
| AK313384.1 | - | 7 - 8 | 1250 | 83 | ||
| BC017033.1 | - | 7 - 8 | 1364 | 3846346 | 83 | |
| BX647400.1 | - | 7 - 8 | 2066 | 83 | ||
| BX647605.1 | - | 7 - 8 | 2104 | 83 | ||
| D78129.1 | - | 5 - 6 | 650 | 83 | ||
| D78130.1 | - | 7 - 8 | 1469 | 83 | ||
More Information
Set Membership: |
|

SQLE
Genomic Map