Assay Details
Gene Symbol: RPL30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK308890.1 | - | 3 - 4 | 239 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200010004.hg.1 | ENST00000495304.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010004.hg.1 | ENST00000495304.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010004.hg.1 | ENST00000509114.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010004.hg.1 | ENST00000509114.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010004.hg.1 | ENST00000509114.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010004.hg.1 | ENST00000509114.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010004.hg.1 | ENST00000598995.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000287038.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000287038.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000287038.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000287038.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000287038.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000287038.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000287038.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000396070.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000396070.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000396070.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000396070.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000396070.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000517489.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000517489.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000517489.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000517489.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000517489.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000517805.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000517805.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000517805.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000518164.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000518164.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000518164.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000518850.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000518850.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000518850.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000518850.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000518850.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000518850.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000518850.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000518850.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000520998.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000520998.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000520998.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000520998.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000520998.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521112.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521112.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521112.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521112.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521291.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521291.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521291.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521291.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521291.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521291.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000521726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000522156.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000522156.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000522156.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000522156.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000523172.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000523172.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000523172.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | ENST00000523172.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | NM_000989.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | NM_000989.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | NM_000989.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | NM_000989.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | NM_000989.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | NM_000989.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011206.hg.1 | NM_000989.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010004.hg.1 | ENST00000495304.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010004.hg.1 | ENST00000495304.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010004.hg.1 | ENST00000509114.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010004.hg.1 | ENST00000509114.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010004.hg.1 | ENST00000509114.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010004.hg.1 | ENST00000509114.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010004.hg.1 | ENST00000598995.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000287038.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000287038.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000287038.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000287038.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000287038.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000287038.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000287038.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000396070.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000396070.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000396070.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000396070.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000396070.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000517489.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000517489.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000517489.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000517489.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000517489.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000517805.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000517805.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000517805.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000518164.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000518164.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000518164.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000518850.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000518850.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000518850.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000518850.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000518850.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000518850.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000518850.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000518850.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000520998.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000520998.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000520998.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000520998.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000520998.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521112.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521112.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521112.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521112.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521291.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521291.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521291.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521291.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521291.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521291.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521534.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521534.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521534.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521534.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521534.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521534.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000521726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000522156.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000522156.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000522156.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000522156.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000523172.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000523172.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000523172.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | ENST00000523172.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | NM_000989.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | NM_000989.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | NM_000989.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | NM_000989.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | NM_000989.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | NM_000989.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800011206.hg.1 | NM_000989.3 | Refseq |
