| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008770.hg.1 |
ENST00000303545.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008770.hg.1 |
ENST00000303545.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008770.hg.1 |
ENST00000303545.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008770.hg.1 |
ENST00000303545.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008770.hg.1 |
ENST00000303545.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008770.hg.1 |
ENST00000303545.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008770.hg.1 |
ENST00000303545.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008770.hg.1 |
ENST00000517684.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008770.hg.1 |
ENST00000517684.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008770.hg.1 |
ENST00000518639.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008770.hg.1 |
ENST00000518639.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008770.hg.1 |
ENST00000518639.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008770.hg.1 |
NM_007218.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008770.hg.1 |
NM_007218.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008770.hg.1 |
NM_007218.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008770.hg.1 |
NM_007218.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008770.hg.1 |
NM_007218.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008770.hg.1 |
NM_007218.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008770.hg.1 |
NM_007218.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008770.hg.1 |
NM_007218.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008770.hg.1 |
ENST00000303545.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008770.hg.1 |
ENST00000303545.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008770.hg.1 |
ENST00000303545.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008770.hg.1 |
ENST00000303545.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008770.hg.1 |
ENST00000303545.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008770.hg.1 |
ENST00000303545.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008770.hg.1 |
ENST00000303545.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008770.hg.1 |
ENST00000517684.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008770.hg.1 |
ENST00000517684.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008770.hg.1 |
ENST00000518639.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008770.hg.1 |
ENST00000518639.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008770.hg.1 |
ENST00000518639.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008770.hg.1 |
NM_007218.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008770.hg.1 |
NM_007218.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008770.hg.1 |
NM_007218.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008770.hg.1 |
NM_007218.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008770.hg.1 |
NM_007218.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008770.hg.1 |
NM_007218.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008770.hg.1 |
NM_007218.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008770.hg.1 |
NM_007218.3 |
Refseq |