Assay Details
| Gene Symbol: FBXO32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242463.1 | NP_001229392.1 | 7 | 4661 | 104 | |
| NM_058229.3 | NP_478136.1 | 9 | 4940 | 104 | ||
| NM_148177.2 | NP_680482.1 | 6 | 4575 | 104 | ||
| GenBank mRNA | AK124390.1 | - | 1 | 405 | 104 | |
| AK125888.1 | - | 1 | 3089 | 104 | ||
More Information
Set Membership: |
|

FBXO32
Genomic Map