Assay Details
| Gene Symbol: HHLA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145095.1 | NP_001138567.1 | 16 | 3900 | 86 | |
| GenBank mRNA | AK022619.1 | - | 1 | 1816 | 86 | |
More Information
Set Membership: |
|

HHLA1
Genomic Map