Assay Details
| Gene Symbol: CACNA1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000718.3 | NP_000709.1 | 17 - 18 | 2298 | 65 | |
| NM_001243812.1 | NP_001230741.1 | 17 - 18 | 2298 | 65 | ||
| XM_011518991.2 | XP_011517293.1 | 17 - 18 | 2265 | 65 | ||
| XM_011518992.2 | XP_011517294.1 | 17 - 18 | 2265 | 65 | ||
| XM_011518993.2 | XP_011517295.1 | 17 - 18 | 2261 | 65 | ||
| XM_011518994.2 | XP_011517296.1 | 17 - 18 | 2264 | 65 | ||
| XM_017015115.1 | XP_016870604.1 | 17 - 18 | 2262 | 65 | ||
| XM_017015116.1 | XP_016870605.1 | 17 - 18 | 2265 | 65 | ||
| XM_017015117.1 | XP_016870606.1 | 17 - 18 | 2264 | 65 | ||
| XM_017015118.1 | XP_016870607.1 | 17 - 18 | 2261 | 65 | ||
| XM_017015119.1 | XP_016870608.1 | 17 - 18 | 2264 | 65 | ||
| XM_017015120.1 | XP_016870609.1 | 17 - 18 | 2261 | 65 | ||
| GenBank mRNA | M94172.1 | - | 17 - 18 | 2298 | 65 | |
| M94173.1 | - | 17 - 18 | 2298 | 65 | ||
More Information
Set Membership: |
|

CACNA1B
Genomic Map