Assay Details
| Gene Symbol: DAPK1-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BQ025444.1 | - | 1 - 2 | 574 | 111 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DAPK1-IT1| Gene Symbol: DAPK1-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BQ025444.1 | - | 1 - 2 | 574 | 111 | |
Set Membership: |
|