Assay Details
| Gene Symbol: SLC24A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005251425.2 | XP_005251482.1 | 1 - 2 | 277 | 77 | |
| XM_017014593.1 | XP_016870082.1 | 1 - 2 | 281 | 77 | ||
| GenBank mRNA | DQ067462.1 | - | 1 - 2 | 78 | 77 | |
| DQ067463.1 | - | 1 - 2 | 278 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005251425.2, XM_017014593.1 transcripts.
Set Membership: |
|

SLC24A2
Genomic Map