Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010637.hg.1 |
ENST00000298743.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010637.hg.1 |
ENST00000298743.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010637.hg.1 |
ENST00000298743.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010637.hg.1 |
ENST00000298743.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010637.hg.1 |
ENST00000298743.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010637.hg.1 |
ENST00000298743.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010637.hg.1 |
ENST00000298743.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010637.hg.1 |
NM_002048.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010637.hg.1 |
NM_002048.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010637.hg.1 |
NM_002048.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010637.hg.1 |
NM_002048.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010637.hg.1 |
NM_002048.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010637.hg.1 |
NM_002048.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010637.hg.1 |
NM_002048.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010637.hg.1 |
ENST00000298743.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010637.hg.1 |
ENST00000298743.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010637.hg.1 |
ENST00000298743.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010637.hg.1 |
ENST00000298743.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010637.hg.1 |
ENST00000298743.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010637.hg.1 |
ENST00000298743.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010637.hg.1 |
ENST00000298743.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010637.hg.1 |
NM_002048.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010637.hg.1 |
NM_002048.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010637.hg.1 |
NM_002048.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010637.hg.1 |
NM_002048.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010637.hg.1 |
NM_002048.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010637.hg.1 |
NM_002048.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010637.hg.1 |
NM_002048.2 |
Refseq |