Assay Details
| Gene Symbol: UBAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016172.2 | NP_057256.2 | 8 - 9 | 1181 | 56 | |
| GenBank mRNA | AF038200.1 | - | 7 - 8 | 746 | 56 | |
| AF068195.1 | - | 7 - 8 | 808 | 56 | ||
| AF176796.1 | - | 8 - 9 | 1151 | 56 | ||
| BC004967.1 | - | 8 - 9 | 1181 | 3535682 | 56 | |
| BC011822.1 | - | 8 - 9 | 1007 | 3688526 | 56 | |
| BX647463.1 | - | 1 - 2 | 5234 | 56 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000465873.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000465873.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000465873.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000465873.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000465873.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000465873.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000471163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000471163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000471163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000471163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000471163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000471163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000471163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000471163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000471163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000478485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000478485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000478485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000478485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000478485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000478485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000478485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000486258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000486258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000486258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000489050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000489050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000489050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000489050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000489050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000489050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | ENST00000489050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000371756.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000465873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000465873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000465873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000465873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000465873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000465873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000471163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000471163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000471163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000471163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000471163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000471163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000471163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000471163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000471163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000478485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000478485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000478485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000478485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000478485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000478485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000478485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000486258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000486258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000486258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000489050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000489050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000489050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000489050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000489050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000489050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | ENST00000489050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | NM_016172.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011959.hg.1 | NM_016172.2 | Refseq |

UBAC1
Genomic Map