Assay Details
| Gene Symbol: PTCH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083602.1 | NP_001077071.1 | 1 - 2 | 498 | 80 | |
| GenBank mRNA | AB189436.1 | - | 1 - 2 | 498 | 80 | |
| BC043542.1 | - | 1 - 2 | 590 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB189436.1, BC043542.1, NM_001083602.1 transcripts.
Set Membership: |
|

PTCH1
Genomic Map