Assay Details
| Gene Symbol: LOC101928722 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011519280.2 | XP_011517582.1 | 5 - 6 | 621 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928722| Gene Symbol: LOC101928722 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011519280.2 | XP_011517582.1 | 5 - 6 | 621 | 64 | |
Set Membership: |
|