Assay Details
| Gene Symbol: UTP14A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017029237.1 | XP_016884726.1 | 8 - 9 | 781 | 90 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000394422.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000394422.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000394422.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000394422.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000394422.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000394422.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000394422.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000394422.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000394422.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000394422.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000394422.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000394422.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000394422.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000394422.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000394422.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000394422.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000394422.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000394422.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000425117.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000425117.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000425117.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000425117.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000425117.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000425117.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000425117.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000425117.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000425117.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000425117.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000425117.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000425117.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000425117.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000425117.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000425117.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000425117.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000425117.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000425117.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000427972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000427972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000427972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000427972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000427972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000427972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000427972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000427972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000498179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000498179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | ENST00000498179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | NM_001166221.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | NM_001166221.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | NM_001166221.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | NM_001166221.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | NM_001166221.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | NM_001166221.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | NM_001166221.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | NM_001166221.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | NM_001166221.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | NM_001166221.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | NM_001166221.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | NM_001166221.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | NM_001166221.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | NM_001166221.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | NM_001166221.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | NM_001166221.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | NM_001166221.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | NM_001166221.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | NM_006649.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | NM_006649.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | NM_006649.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | NM_006649.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | NM_006649.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | NM_006649.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | NM_006649.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | NM_006649.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | NM_006649.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | NM_006649.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | NM_006649.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | NM_006649.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | NM_006649.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | NM_006649.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | NM_006649.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | NM_006649.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | NM_006649.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | NM_006649.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008392.hg.1 | NM_006649.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000394422.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000394422.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000394422.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000394422.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000394422.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000394422.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000394422.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000394422.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000394422.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000394422.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000394422.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000394422.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000394422.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000394422.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000394422.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000394422.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000394422.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000394422.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000425117.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000425117.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000425117.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000425117.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000425117.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000425117.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000425117.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000425117.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000425117.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000425117.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000425117.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000425117.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000425117.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000425117.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000425117.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000425117.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000425117.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000425117.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000427972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000427972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000427972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000427972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000427972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000427972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000427972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000427972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000498179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000498179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | ENST00000498179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | NM_001166221.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | NM_001166221.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | NM_001166221.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | NM_001166221.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | NM_001166221.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | NM_001166221.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | NM_001166221.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | NM_001166221.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | NM_001166221.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | NM_001166221.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | NM_001166221.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | NM_001166221.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | NM_001166221.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | NM_001166221.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | NM_001166221.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | NM_001166221.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | NM_001166221.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | NM_001166221.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | NM_006649.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | NM_006649.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | NM_006649.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | NM_006649.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | NM_006649.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | NM_006649.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | NM_006649.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | NM_006649.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | NM_006649.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | NM_006649.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | NM_006649.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | NM_006649.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | NM_006649.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | NM_006649.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | NM_006649.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | NM_006649.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | NM_006649.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | NM_006649.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008392.hg.1 | NM_006649.3 | Refseq |

UTP14A
Genomic Map