Assay Details
| Gene Symbol: STS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320750.1 | NP_001307679.1 | 2 - 3 | 481 | 129 | |
| NM_001320751.1 | NP_001307680.1 | 3 - 4 | 475 | 129 | ||
| NM_001320752.1 | NP_001307681.1 | 2 - 3 | 321 | 129 | ||
| GenBank mRNA | DA327298.1 | - | 2 - 3 | 284 | 129 | |
| DA406759.1 | - | 2 - 3 | 322 | 129 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001320750.1, NM_001320751.1, NM_001320752.1 transcripts.
Set Membership: |
|

STS
Genomic Map