Assay Details
| Gene Symbol: ZDHHC9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011531348.2 | XP_011529650.1 | 5 - 6 | 916 | 85 | |
More Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000406492.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000406492.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000406492.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000406492.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000406492.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000406492.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000406492.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000406492.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000433917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000433917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000433917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000433917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000433917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000433917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000433917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000491039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000491039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000406492.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000406492.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000406492.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000406492.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000406492.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000406492.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000406492.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000406492.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000433917.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000433917.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000433917.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000433917.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000433917.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000433917.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000433917.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000491039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000491039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |

ZDHHC9
Genomic Map