Assay Details
| Gene Symbol: HNRNPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135167.1 | 10 - 11 | 1152 | 128 | ||
| GenBank mRNA | BC070315.1 | - | 10 - 11 | 1094 | 4101267 | 128 |
| X79536.1 | - | 10 - 11 | 1060 | 128 | ||
More Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splicing, via spliceosome
mRNA processing
RNA export from nucleus
fibroblast growth factor receptor signaling pathway
gene expression
viral process
negative regulation of telomere maintenance via telomerase
positive regulation of telomere maintenance via telomerase
mRNA transport
nuclear export
nuclear import
nucleotide binding
single-stranded DNA binding
RNA binding
single-stranded RNA binding
protein binding
poly(A) RNA binding
telomeric repeat-containing RNA binding
G-rich strand telomeric DNA binding
mRNA splicing, via spliceosome
mRNA processing
RNA export from nucleus
fibroblast growth factor receptor signaling pathway
gene expression
viral process
negative regulation of telomere maintenance via telomerase
positive regulation of telomere maintenance via telomerase
mRNA transport
nuclear export
nuclear import
nucleotide binding
single-stranded DNA binding
RNA binding
single-stranded RNA binding
protein binding
poly(A) RNA binding
telomeric repeat-containing RNA binding
G-rich strand telomeric DNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000330752.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000330752.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000330752.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000330752.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000330752.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000330752.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000330752.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000330752.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000330752.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000330752.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000330752.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000330752.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000330752.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000330752.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000340913.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000340913.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000340913.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000340913.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000340913.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000340913.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000340913.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000340913.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000340913.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000340913.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000340913.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000340913.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000340913.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000340913.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000340913.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000340913.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000340913.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000546500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000546500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000546500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000546500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000546500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000546500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000546500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000546500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000546500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000546500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000546500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000546500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000546500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000546500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000546500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000546500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000546500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000546500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000546500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000546500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000546500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000546500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000546500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000546500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000546500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000546500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000546500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547566.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547566.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547566.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547566.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547566.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547566.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547566.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547566.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547566.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547566.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547566.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547566.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547566.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547566.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547566.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547566.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547708.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547708.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547708.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547708.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547708.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547708.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000547870.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000548688.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000548688.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000548688.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000548688.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000548688.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000548688.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000548688.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000548688.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000550482.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000550482.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000550482.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000550482.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000550482.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000550482.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000550482.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000550482.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000550482.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000550482.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000550482.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000550994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000550994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000550994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000550994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000550994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000550994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000551665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000551665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000551665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000551665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000551665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000551679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000551679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000551679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000551679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000551679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000551702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000551702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000551702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000551702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000551702.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000551803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000551803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000551803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | ENST00000551803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | NM_002136.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | NM_002136.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | NM_002136.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | NM_002136.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | NM_002136.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | NM_002136.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | NM_002136.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | NM_002136.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | NM_002136.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | NM_002136.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | NM_002136.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | NM_002136.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | NM_002136.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | NM_002136.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | NM_002136.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | NM_002136.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | NM_002136.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | NM_002136.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | NM_031157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | NM_031157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | NM_031157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | NM_031157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | NM_031157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | NM_031157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | NM_031157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | NM_031157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | NM_031157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | NM_031157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | NM_031157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | NM_031157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | NM_031157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | NM_031157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | NM_031157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | NM_031157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | NM_031157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | NM_031157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007758.hg.1 | NM_031157.3 | Refseq |

HNRNPA1
Genomic Map