| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009355.hg.1 |
ENST00000536342.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009355.hg.1 |
ENST00000536342.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009355.hg.1 |
ENST00000536342.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009355.hg.1 |
ENST00000536342.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009355.hg.1 |
ENST00000614177.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009355.hg.1 |
ENST00000614177.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009355.hg.1 |
ENST00000614177.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009355.hg.1 |
ENST00000614177.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009355.hg.1 |
ENST00000614177.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009355.hg.1 |
NR_033987.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009355.hg.1 |
NR_033987.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009355.hg.1 |
NR_033987.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009355.hg.1 |
NR_033987.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009355.hg.1 |
NR_033987.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200009355.hg.1 |
NR_033987.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009355.hg.1 |
ENST00000536342.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009355.hg.1 |
ENST00000536342.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009355.hg.1 |
ENST00000536342.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009355.hg.1 |
ENST00000536342.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009355.hg.1 |
ENST00000614177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009355.hg.1 |
ENST00000614177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009355.hg.1 |
ENST00000614177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009355.hg.1 |
ENST00000614177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009355.hg.1 |
ENST00000614177.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200009355.hg.1 |
NR_033987.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009355.hg.1 |
NR_033987.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009355.hg.1 |
NR_033987.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009355.hg.1 |
NR_033987.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009355.hg.1 |
NR_033987.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200009355.hg.1 |
NR_033987.1 |
Refseq |