Assay Details
| Gene Symbol: PARPBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319988.1 | NP_001306917.1 | 7 - 8 | 1170 | 165 | |
| NM_001319993.1 | NP_001306922.1 | 7 - 8 | 1047 | 165 | ||
| NM_001319994.1 | NP_001306923.1 | 6 - 7 | 934 | 165 | ||
| NM_017915.4 | NP_060385.3 | 6 - 7 | 939 | 165 | ||
| XM_011538509.1 | XP_011536811.1 | 5 - 6 | 705 | 165 | ||
| XM_011538514.2 | XP_011536816.1 | 5 - 6 | 752 | 165 | ||
| XM_011538515.1 | XP_011536817.1 | 4 - 5 | 545 | 165 | ||
| XM_017019534.1 | XP_016875023.1 | 5 - 6 | 768 | 165 | ||
| XM_017019535.1 | XP_016875024.1 | 5 - 6 | 778 | 165 | ||
| XM_017019536.1 | XP_016875025.1 | 6 - 7 | 939 | 165 | ||
| XM_017019538.1 | XP_016875027.1 | 6 - 7 | 939 | 165 | ||
| XM_017019541.1 | XP_016875030.1 | 6 - 7 | 939 | 165 | ||
| XM_017019542.1 | XP_016875031.1 | 6 - 7 | 939 | 165 | ||
| GenBank mRNA | AK000648.1 | - | 6 - 7 | 858 | 165 | |
| AK302724.1 | - | 7 - 8 | 1156 | 165 | ||
| AK303571.1 | - | 6 - 7 | 865 | 165 | ||
| BC030962.1 | - | 7 - 8 | 991 | 165 | ||
| BC070270.1 | - | 6 - 7 | 895 | 165 | ||
| BC098313.1 | - | 3 - 4 | 329 | 40025466 | 165 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC098313.1 transcripts.
Set Membership: |
|

PARPBP
Genomic Map