Assay Details
| Gene Symbol: PARPBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319988.1 | NP_001306917.1 | 8 - 9 | 1355 | 85 | |
| NM_001319993.1 | NP_001306922.1 | 8 - 9 | 1232 | 85 | ||
| NM_001319994.1 | NP_001306923.1 | 7 - 8 | 1119 | 85 | ||
| NM_017915.4 | NP_060385.3 | 7 - 8 | 1124 | 85 | ||
| XM_011538509.1 | XP_011536811.1 | 6 - 7 | 890 | 85 | ||
| XM_011538514.2 | XP_011536816.1 | 6 - 7 | 937 | 85 | ||
| XM_011538515.1 | XP_011536817.1 | 5 - 6 | 730 | 85 | ||
| XM_017019534.1 | XP_016875023.1 | 6 - 7 | 953 | 85 | ||
| XM_017019535.1 | XP_016875024.1 | 6 - 7 | 963 | 85 | ||
| XM_017019538.1 | XP_016875027.1 | 7 - 8 | 1124 | 85 | ||
| GenBank mRNA | AK000648.1 | - | 7 - 8 | 1043 | 85 | |
| AK302724.1 | - | 8 - 9 | 1341 | 85 | ||
| AK303571.1 | - | 7 - 8 | 1050 | 85 | ||
| BC030962.1 | - | 8 - 9 | 1176 | 85 | ||
| BC070270.1 | - | 7 - 8 | 1080 | 85 | ||
| BC098313.1 | - | 4 - 5 | 514 | 40025466 | 85 | |
More Information
Set Membership: |
|

PARPBP
Genomic Map