Assay Details
| Gene Symbol: PARPBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319988.1 | NP_001306917.1 | 9 - 10 | 1538 | 89 | |
| NM_001319993.1 | NP_001306922.1 | 9 - 10 | 1415 | 89 | ||
| NM_001319994.1 | NP_001306923.1 | 8 - 9 | 1302 | 89 | ||
| NM_017915.4 | NP_060385.3 | 8 - 9 | 1307 | 89 | ||
| XM_011538509.1 | XP_011536811.1 | 7 - 8 | 1073 | 89 | ||
| XM_011538514.2 | XP_011536816.1 | 7 - 8 | 1120 | 89 | ||
| XM_011538515.1 | XP_011536817.1 | 6 - 7 | 913 | 89 | ||
| XM_017019534.1 | XP_016875023.1 | 7 - 8 | 1136 | 89 | ||
| XM_017019535.1 | XP_016875024.1 | 7 - 8 | 1146 | 89 | ||
| XM_017019538.1 | XP_016875027.1 | 8 - 9 | 1307 | 89 | ||
| GenBank mRNA | AK000648.1 | - | 8 - 9 | 1226 | 89 | |
| AK302724.1 | - | 9 - 10 | 1524 | 89 | ||
| AK303571.1 | - | 8 - 9 | 1233 | 89 | ||
| BC030962.1 | - | 9 - 10 | 1359 | 89 | ||
| BC070270.1 | - | 8 - 9 | 1263 | 89 | ||
| BC098313.1 | - | 5 - 6 | 697 | 40025466 | 89 | |
More Information
Set Membership: |
|

PARPBP
Genomic Map